Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Trioxide, Crystal, ACS, 98%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
1333-82-0
1333-82-0
Grade ACS
CAS Max % 1
Aluminum (Al) 0.0002
Barium (Ba) 0.0001
Percent Purity 0.98
Chloride 5e-05
Insoluble Matter 0.0001
Iron (Fe) 0.0002
Nitrate 0.0005
Packaging Amber Glass Bottle
Sodium (Na) 0.002
Sulfate 5e-05
Quantity 125 g
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.