Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Trioxide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

1333-82-0
CrO3
99.993
WGLPBDUCMAPZCE-UHFFFAOYSA-N
14915
CHEBI:48240

High quality chemicals for laboratory and specialized industrial use

CAS 1333-82-0
Molecular Formula CrO3
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
SMILES O=[Cr](=O)=O
IUPAC Name trioxochromium
Molecular Weight (g/mol) 99.993
PubChem CID 14915
ChEBI CHEBI:48240
Formula Weight 99.99
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.70kg
Quantity 500g
Chemical Name or Material Chromium Trioxide
Grade Baker Analyzed™ ACS Reagent
Show More Show Less