Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Trioxide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle
1333-82-0
CrO3
99.993
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide,chromium vi oxide,monochromium trioxide,chromtrioxid,chromic trioxide,chromsaeureanhydrid,anhydride chromique,chromium 6+ oxide,chromium vi trioxide,chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O
1333-82-0
CrO3
99.993
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide,chromium vi oxide,monochromium trioxide,chromtrioxid,chromic trioxide,chromsaeureanhydrid,anhydride chromique,chromium 6+ oxide,chromium vi trioxide,chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 99.99
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.70kg
Quantity 125g
Chemical Name or Material Chromium Trioxide