Chromium Trioxide (Flakes/Technical), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 2.5kg

 View more versions of this product

Catalog No. A98-212

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 1333-82-0
    Physical Form Solid
    Molecular Formula CrO3
    Formula Weight 99.99
    MDL Number MFCD00010952
    Synonym Chromic Acid Anhydride
    Chemical Name or Material Chromium Trioxide
    Grade Technical Grade

    Specifications

    Formula Weight 99.99
    Melting Point 196°C
    Quantity 2.5kg
    DOT Information DOT Class 5.1, : Oxidizer
    Physical Form Solid
    Color Reddish-violet
    pH 1
    Density 3.4
    Identification Pass Test
    Packaging Glass Bottle
    MDL Number MFCD00010952
    ChemAlert Storage Symbol Yellow
    Health Hazard 2 DANGER!
    Molecular Formula CrO3
    CAS 1333-82-0
    Synonym Chromic Acid Anhydride
    Health Hazard 3 Emergency Overview
    Explosive when mixed with combustible material. May be fatal if inhaled. Causes severe burns by all exposure routes. Toxic in contact with skin and if swallowed. May cause sensitization by inhalation and skin contact. Danger of serious damage to health by prolonged exposure. May cause heritable genetic damage. Possible risk of impaired fertility. Hygroscopic. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Wear personal protective equipment. Use only under a chemical fume hood. Use only under a chemical fume hood. Keep away from clothing and other combustible materials.

    NFPA

    Health:3
    Flammability:0
    Instability:1
    Chemical Name or Material Chromium Trioxide
    Grade Technical Grade
     

    DANGER!

    Emergency Overview
    Explosive when mixed with combustible material. May be fatal if inhaled. Causes severe burns by all exposure routes. Toxic in contact with skin and if swallowed. May cause sensitization by inhalation and skin contact. Danger of serious damage to health by prolonged exposure. May cause heritable genetic damage. Possible risk of impaired fertility. Hygroscopic. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Wear personal protective equipment. Use only under a chemical fume hood. Use only under a chemical fume hood. Keep away from clothing and other combustible materials.

    NFPA

    Health:3
    Flammability:0
    Instability:1

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit