Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(VI) oxide, 99%, Thermo Scientific Chemicals

Catalog Number AC196610250
Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
1333-82-0
CrO3
99.99
MFCD00010952
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O
1333-82-0
CrO3
99.99
MFCD00010952
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 196.0°C
Color Purple to Red
Physical Form Crystals or Granules
Quantity 25 g
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula CrO3
Fieser 01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
Merck Index 15, 2239
Solubility Information Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids
Formula Weight 99.99
Percent Purity 99.5%
Packaging Glass bottle
Specific Gravity 2.7
Density 2.7000g/mL
Chemical Name or Material Chromium(VI) oxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
EINECSNumber 215-607-8

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit