Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(VI) oxide, 99.5%, ACROS Organics™

Click to view available options
Quantity:
25g
250g
1kg
2.5kg
Packaging:
Glass bottle
1333-82-0
CrO3
99.99
MFCD00010952
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide,chromium vi oxide,monochromium trioxide,chromtrioxid,chromic trioxide,chromsaeureanhydrid,anhydride chromique,chromium 6+ oxide,chromium vi trioxide,chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O
1333-82-0
CrO3
99.99
MFCD00010952
WGLPBDUCMAPZCE-UHFFFAOYSA-N
chromium trioxide,chromium vi oxide,monochromium trioxide,chromtrioxid,chromic trioxide,chromsaeureanhydrid,anhydride chromique,chromium 6+ oxide,chromium vi trioxide,chromium oxide
14915
CHEBI:48240
trioxochromium
O=[Cr](=O)=O
Oxidation reagent
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula CrO3
Fieser 01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
Merck Index 15, 2239
Solubility Information Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids
Formula Weight 99.99
Physical Form Crystals or Granules
Percent Purity 99.5%
Density 2.7000g/mL
Packaging Glass bottle
Solubility Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids
Specific Gravity 2.7
Sulfate 500ppm max.
Color Purple to Red
Melting Point 196.0°C
Quantity 250g
Chemical Name or Material Chromium(VI) oxide
Show More Show Less

GHS H Statement
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 215-607-8