Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium(VI) oxide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Melting Point | 196.0°C |
Color | Purple to Red |
Physical Form | Crystals or Granules |
Quantity | 25 g |
Assay Percent Range | 99.4% min. (Iodometry) |
Linear Formula | CrO3 |
Fieser | 01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60 |
Merck Index | 15, 2239 |
Solubility Information | Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids |
Formula Weight | 99.99 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse |
EINECSNumber | 215-607-8 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction