Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Chromogranin A Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS324PABX

 View more versions of this product

Catalog No. MS324PABX


Add to cart

Description

Description

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.

Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: LK2H10

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: PC-12 cells, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Chromogranin A Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgG2a kappa
IVD
Primary
Monoclonal
Immunohistochemistry (Paraffin), Western Blot
LK2H10
Unconjugated
Concentrated
Human pheochromocytoma
200μL
Oncogenesis
Primary
Human, Non-human Primate, Porcine, Rabbit, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit