Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin A Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.

Supplier:  Epredia MS382P1

 View more versions of this product

Catalog No. MS382P1


Add to Cart

Description

Description

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Ab-3 cocktail is especially designed for sensitive detection of Chromogranin A in formalin-fixed, paraffin-embedded tissues.

Host Species: Mouse

Clone: LK2H10 + PHE5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Monkey. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic in endocrine cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Chromogranin A Ab-3
Monoclonal
200μg/mL
Mouse
500 μL
Oncogenesis
Human, Non-human Primate
IgG1
Immunohistochemistry (Paraffin)
LK2H10 + PHE5
Unconjugated
Human pheochromocytoma
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit