Learn More
Chromogranin A Antibody (OTI2E12), DyLight 550, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270410R
Catalog No. NBP270410R
Add to Cart
Description
Description
Chromogranin A Monoclonal specifically detects Chromogranin A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Chromogranin A
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
CHGA
Full length recombinant protein of human Chromogranin A (NP_001266) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2E12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I
Mouse
Affinity Purified
Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers
1113.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions