Learn More
Chromogranin A Mouse anti-Human, Mouse, Rat, Porcine, Alexa Fluor™ 750, Clone: LK2H10+ PHE5+ CGA/414, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234674AF750
Catalog No. NP234674A70
Add to cart
Description
Description
Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human,Mouse,Rat,Porcine,Primate samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
Chromogranin A
LK2H10 + PHE5 + CGA/414
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I
Mouse
IgG1 Kappa/IgG1 Kappa/IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CHGA
Human pheochromocytoma (LK2H10 & PHE5) & recombinant human chromogranin A protein (CGA414)
0.1 mL
Primary
1113
Human, Mouse, Rat, Porcine, Primate