Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin A Rabbit anti-Human, Rat, Alexa Fluor™ 750, Clone: CHGA/1815R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP254363AF750

Catalog No. NP24363A70


Add to cart

Description

Description

Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human,Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications

Specifications

Chromogranin A
CHGA/1815R
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Human, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CHGA
Recombinant full-length human CHGA protein
0.1 mL
Primary
1113
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit