Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Chromogranin A Rabbit Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ Chromogranin A, Rabbit Monoclonal Antibody.

Supplier:  Epredia™ RM9112R7

 View more versions of this product

Catalog No. RM9112R7


Add to Cart

Description

Description

Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Rabbit

Clone: SP12

Isotype: IgG

Species Reactivity: Human. Others not known

Epitope: Not determined

Immunogen: Recombinant protein encoding human chromogranin A

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Chromogranin A
Monoclonal
Unconjugated
Recombinant protein encoding human chromogranin A
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
SP12
Rabbit
7 mL
Oncogenesis
Human
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit