Learn More
Chromogranin C Mouse anti-Rat, DyLight™ 650, Clone: SGII 6B1/3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP276658C
Catalog No. NBP276658C
Add to cart
Description
Description
Chromogranin C Monoclonal antibody specifically detects Chromogranin C in Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen).
Specifications
Specifications
Chromogranin C
SGII 6B1/3
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
SCG2
Peptide from rat SGII (C terminus of p18: KEENSRENPF)
0.1 mL
Primary
7857