Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromogranin C Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19162820UL

 View more versions of this product

Catalog No. NBP19162820


Add to cart

Description

Description

Chromogranin C Polyclonal antibody specifically detects Antigen in Rat samples. It is validated for Western Blotting.
Specifications

Specifications

Chromogranin C
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
SCG2
Synthetic peptide directed towards SCG2. Peptide sequence DIDRPDMFQSKTLSKGGYPKAPGRGMMEALPDGLSVEDILNVLGMENVAN.
Immunogen affinity purified
RUO
Primary
7857
Western Blot
Western Blot 1:1000
NP_073160
CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit