Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromone-3-carboxaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17422-74-1
C10H6O3
174.155
MFCD00014667
FSMYWBQIMDSGQP-UHFFFAOYSA-N
chromone-3-carboxaldehyde,3-formylchromone,4-oxo-4h-chromene-3-carbaldehyde,3-chromonecarbaldehyde,4-oxo-4h-1-benzopyran-3-carboxaldehyde,3-formylchromones,4h-1-benzopyran-3-carboxaldehyde, 4-oxo,chembl86905,3-formyl-4-oxo-4h-chromene,chromone-3-carbaldehyde
87112
4-oxochromene-3-carbaldehyde
C1=CC=C2C(=C1)C(=O)C(=CO2)C=O
17422-74-1
C10H6O3
174.155
MFCD00014667
FSMYWBQIMDSGQP-UHFFFAOYSA-N
chromone-3-carboxaldehyde,3-formylchromone,4-oxo-4h-chromene-3-carbaldehyde,3-chromonecarbaldehyde,4-oxo-4h-1-benzopyran-3-carboxaldehyde,3-formylchromones,4h-1-benzopyran-3-carboxaldehyde, 4-oxo,chembl86905,3-formyl-4-oxo-4h-chromene,chromone-3-carbaldehyde
87112
4-oxochromene-3-carbaldehyde
C1=CC=C2C(=C1)C(=O)C(=CO2)C=O
Packaging Glass bottle
Quantity 5g
Color Orange
Melting Point 151°C to 155°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Formula Weight 174.16
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Chromone-3-carboxaldehyde

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

AIR SENSITIVE

EINECSNumber : 241-451-5