Learn More
Alfa Aesar™ Chromostatin, Bovine
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66182MCR
 View more versions of this product
Catalog No. AAJ66182MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1954.00
Bovine
Solid
RUO
Chromostatin-20
HPLC
1mg
Ser-Asp-Glu-Asp-Ser-Asp-Gly-Asp-Arg-Pro-Gln-Ala-Ser-Pro-Gly-Leu-Gly-Pro-Gly-Pro
-30° to -10°C
Chromostatin, bovine