Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chromotrope FB

Click to view available options
Quantity:
10g
50g
250g
3567-69-9
C20H12N2Na2O7S2
502.423
MFCD00003978
OJNIRNOPCQQNRY-UHFFFAOYSA-L
Acid Red 14; C.I. 14720
44135716
disodium;4-oxo-3-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-1-sulfonate
C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])NN=C3C=C(C4=CC=CC=C4C3=O)S(=O)(=O)[O-].[Na+].[Na+]
3567-69-9
C20H12N2Na2O7S2
502.423
MFCD00003978
OJNIRNOPCQQNRY-UHFFFAOYSA-L
Acid Red 14; C.I. 14720
44135716
disodium;4-oxo-3-[(4-sulfonatonaphthalen-1-yl)hydrazinylidene]naphthalene-1-sulfonate
C1=CC=C2C(=C1)C(=CC=C2S(=O)(=O)[O-])NN=C3C=C(C4=CC=CC=C4C3=O)S(=O)(=O)[O-].[Na+].[Na+]
Formula Weight 502.42
Quantity 50g
Chemical Name or Material Chromotrope FB

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA