Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromotropic Acid, Disodium Salt, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
Packaging:
Wide Mouth Amber Glass Bottle
5808-22-0
C10H12Na2O10S2
402.296
BAXLVCPBGWZLAU-UHFFFAOYSA-N
chromotropic acid sodium salt,4,5-dihydroxy-2,7-naphthalenedisulfonate,di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid,4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g
124202444
4,5-dihydroxynaphthalene-2,7-disulfonic acid;sodium;dihydrate
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)O)O)S(=O)(=O)O.O.O.[Na].[Na]
5808-22-0
C10H12Na2O10S2
402.296
BAXLVCPBGWZLAU-UHFFFAOYSA-N
chromotropic acid sodium salt,4,5-dihydroxy-2,7-naphthalenedisulfonate,di sodium; 4,5-dihydroxy-7-sulfo-naphthalene-2-sulfonic acid,4,5-dihydroxynaphthalene-2,7-disulfonic acid disodium salt dihydrate, acs 5g
124202444
4,5-dihydroxynaphthalene-2,7-disulfonic acid;sodium;dihydrate
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)O)O)S(=O)(=O)O.O.O.[Na].[Na]

High quality chemicals for laboratory and specialized industrial use

Quantity 25g
Packaging Wide Mouth Amber Glass Bottle
Formula Weight 400.29
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Chromotropic Acid, Disodium Salt, Dihydrate