Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromotropic Acid TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle

C-286, 5808-22-0

Spectrum™ Chromotropic Acid TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 5808-22-0,7664-93-9
CAS Max % 0.1%,20%
Molecular Formula C10H6O8S2
UN Number UN2796
InChI Key HLVXFWDLRHCZEI-UHFFFAOYSA-L
SMILES OC1=CC(=CC2=CC(=CC(O)=C12)S([O-])(=O)=O)S([O-])(=O)=O
IUPAC Name 4,5-dihydroxynaphthalene-2,7-disulfonate
Molecular Weight (g/mol) 318.27
Packaging Amber Glass Bottle
Quantity 100 mL