Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Click to view available options
Packaging:
Amber Glass
Quantity:
1 L
4 L
25 L
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
75-05-8
C2H3N
41.05
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitril, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP.

Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
Absorbance absorbance at 195 nm = < 0.1, absorbance at 200 nm = < 0.05, absorbance at 210 nm = < 0.02, absorbance at 220 nm = < 0.01
CAS Min % 0.9995
Absorbance Conditions 190 nm,195 nm,200 nm,205 nm,210 nm,220 nm,254 nm
Absorbance Scale AU
Additional Information ≤ 1 ppb
UN Number 1648
Formula Weight 41.05
Physical Form Liquid
Percent Purity 99.95%
Grade Chromplete
Density 0.775 - 0.78
Gradient Analysis at 205 nm, 254 nm
HPLC Gradient Suitability ≤ 5.4 mA.U.,≤ 1.4 mA.U.
Packaging Amber Glass
Pesticide Residue Analysis ≤ 10 ng/L
Refractive Index 1.3405 – 1.3425
Residue after Ignition Must be 'Pass'
Residue after Evaporation ≤ 0.8 ppm
Titratable Acid ≤ 0.008 mEq/g
Titratable Base ≤ 0.0006 mEq/g
Water ≤ 0.01 %
Boiling Point 180°F
Color ≤ 5 APHA
Melting Point -51°F
Quantity 1 L
Assay Minimum, GC-FID
Show More Show Less

Causes serious eye irritation, Highly flammable liquid and vapor, Harmful if swallowed, in contact with skin or if inhaled

Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Acute toxicity (category 4)

ShelfLife : 1825 days

EINECSNumber : 200-835-2

RTECSNumber : AL7700000

storageNote1 : In sealed containers

Recommended Storage : Room temperature flammable storage

RUO – Research Use Only