Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

Catalog Number p-8776008
Click to view available options
Quantity:
1 L
4 L
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Methanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY
Melting Point -98°C
Absorbance ≤1.00, ≤ 0.23, ≤ 0.11, ≤ 0.01
Color ≤ 5 APHA
CAS Min % 0.999
Density 0.775 – 0.780
Boiling Point 148.5°F
Quantity 1 L
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤ 1.0 ppm
Titratable Acid ≤0.0003 meq/g
Titratable Base ≤0.0002 meq/g
UN Number 1230
Water ≤0.07%
Formula Weight 32.04
Percent Purity 99.9%
Grade Chromplete
Assay Minimum, GC-FID
Chemical Name or Material Methanol
Show More Show Less
Hazard Category Flammable liquids (category 2), Specific target organ toxicity after single exposure (category 1), Acute toxicity (category 3)
Hazard Statement Highly flammable liquid and vapor, Causes damage to organs, Toxic if swallowed, in contact with skin or if inhaled
ShelfLife 1825 days
EINECSNumber 200-659-6
RTECSNumber PC1400000
storageNote1 In sealed containers
Recommended Storage Room temperature flammable storage

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit