Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromplete™ Water, HPLC/GC, and Spectrometry, Thermo Scientific™

Catalog Number p-8776009
Click to view available options
Quantity:
1 L
4 L
7732-18-5
H2O
18.02
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
water
O
7732-18-5
H2O
18.02
XLYOFNOQVPJJNP-UHFFFAOYSA-N
dihydrogen oxide, dihydrogen monoxide
962
CHEBI:15377
water
O

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Water is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

Chromplete Water is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY
Absorbance ≤ 0.01, ≤ 0.005
pH 5.5-7.0
Color ≤5 APHA
Melting Point 0°C
CAS Min % 0.99
Boiling Point 100°C
Formula Weight 18.01
Grade Chromplete
Packaging Amber Glass
Residue after Evaporation ≤ 1.0 ppm
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
ShelfLife 1825 Days
EINECSNumber 231-791-2
RTECSNumber ZC0110000
storageNote1 At room temperature in sealed containers
Recommended Storage Room Temperature

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit