Alfa Aesar™ Chromyl chloride, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4051704

Quantity 2g

 View more versions of this product

Catalog No. AA4051704

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14977-61-8
    Formula Weight 154.9
    MDL Number MFCD00010951
    UN Number UN1758
    Merck Index 14,2248
    Synonym Chromium oxychloride; Chromium(VI) dichloride dioxide
    Chemical Name or Material Chromyl chloride

    Specifications

    Formula Weight 154.9
    Health Hazard 2 GHS H Statement
    H272-H340-H350-H314-H318-H317
    May intensify fire
    oxidizer.
    May cause genetic defects.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 1.911g/mL
    RTECS Number GB5775000
    MDL Number MFCD00010951
    UN Number UN1758
    Health Hazard 1 Danger
    Quantity 2g
    DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CHROMIUM OXYCHLORIDE
    TSCA TSCA
    Physical Form Liquid
    CAS 14977-61-8
    Merck Index 14,2248
    Melting Point −96.5°C
    Boiling Point 117°C
    Synonym Chromium oxychloride; Chromium(VI) dichloride dioxide
    Odor Pungent, makes eyes water
    Sensitivity Moisture Sensitive
    Chemical Name or Material Chromyl chloride
    Assay 99.99%, (metals basis)

    Description

    • Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins

    GHS H Statement
    H272-H340-H350-H314-H318-H317
    May intensify fire
    oxidizer.
    May cause genetic defects.
    May cause cancer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit