Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chrysophenine sodium salt, Thermo Scientific Chemicals

Catalog Number aaj6499918
Click to view available options
Quantity:
50 g
2870-32-8
C30H26N4Na2O8S2
680.66
MFCD00007488
YQMJDPHTMKUEHG-RPKDOIGLSA-L
chrysophenine g, chrysophenine j, chrysophenine y, aurophenine o, chrysophenine gp, chrysophenine ns, direct yellow c, benzo yellow lg, erie yellow y, chrysophenine esp
54603156
sodium;5-[(4-ethoxyphenyl)diazenyl]-2-[(E)-2-[4-[(4-ethoxyphenyl)diazenyl]-2-sulfophenyl]ethenyl]benzenesulfonic acid
[Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(\C=C/C2=CC=C(C=C2S([O-])(=O)=O)N=NC2=CC=C(OCC)C=C2)C(=C1)S([O-])(=O)=O
2870-32-8
C30H26N4Na2O8S2
680.66
MFCD00007488
YQMJDPHTMKUEHG-RPKDOIGLSA-L
chrysophenine g, chrysophenine j, chrysophenine y, aurophenine o, chrysophenine gp, chrysophenine ns, direct yellow c, benzo yellow lg, erie yellow y, chrysophenine esp
54603156
sodium;5-[(4-ethoxyphenyl)diazenyl]-2-[(E)-2-[4-[(4-ethoxyphenyl)diazenyl]-2-sulfophenyl]ethenyl]benzenesulfonic acid
[Na+].[Na+].CCOC1=CC=C(C=C1)N=NC1=CC=C(\C=C/C2=CC=C(C=C2S([O-])(=O)=O)N=NC2=CC=C(OCC)C=C2)C(=C1)S([O-])(=O)=O

A diazo stilbene dye

A diazo stilbene dye

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Orange to Orange-Yellow
Quantity 50 g
Formula Weight 680.66
Physical Form Powder
Chemical Name or Material Chrysophenine sodium salt
EINECSNumber 220-698-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit