Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHST12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549399

Catalog No. PA549399

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the transfer of sulfate to position 4 of the N-acetylgalactosamine (GalNAc) residue of chondroitin and desulfated dermatan sulfate. Chondroitin sulfate constitutes the predomit proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices. Activity toward partially desulfated dermatan sulfate is however lower. Does not form 4, 6-di-O-sulfated GalNAc when chondroitin sulfate C is used as an acceptor.
Specifications

Specifications

CHST12
Polyclonal
PBS with 0.09% sodium azide
Q9NRB3
Carbohydrate sulfotransferase 12, Chondroitin 4-O-sulfotransferase 2, Chondroitin 4-sulfotransferase 2, C4ST-2, C4ST2, Sulfotransferase Hlo, CHST12
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55501
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CHST12
Liquid
CHST12
KLH conjugated synthetic peptide between 72-106 amino acids from the N-terminal region of human CHST12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit