Learn More
CHST13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 201411AP
Catalog No. 50-173-2727
Add to cart
Description
Description
The antibody is specific to CHST13.
The protein encoded by this gene belongs to the sulfotransferase 2 family. It is localized to the Golgi membrane, and catalyzes the transfer of sulfate to the C4 hydroxyl of beta-1, 4-linked N-acetylgalactosamine (GalNAc) flanked by glucuronic acid residue in chondroitin. Chondroitin sulfate constitutes the predomit proteoglycan present in cartilage and is distributed on the surfaces of many cells and extracellular matrices.
Specifications
Specifications
CHST13
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NET6
C4ST 3, C4ST3, CHST13
Rabbit
IgG
150 μL
-20°C
Primary
166012, 500257, 71797
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CHST13
Liquid
Chst13
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat