Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CHST15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531727
Description
Recommended positive controls: Raji. Predicted reactivity: Mouse (94%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GALNAC4S-6ST is a sulfotransferase that transfers sulfate from 3'-phosphoadenosine 5'-phosphosulfate (PAPS) to the C-6 hydroxyl group of the GalNAc 4-sulfate residue of chondroitin sulfate A and forms chondroitin sulfate E containing GlcA-GalNAc(4, 6-SO(4)) repeating units. It also transfers sulfate to a unique non-reducing terminal sequence, GalNAc(4SO4)-GlcA(2SO4)-GalNAc(6SO4), to yield a highly sulfated structure similar to the structure found in thrombomodulin chondroitin sulfate. GALNAC4S-6ST may also act as a B-cell receptor involved in BCR ligation-mediated early activation that mediate regulatory signals key to B-cell development and/or regulation of B-cell-specific RAG expression; however such results are unclear in vivo.
Specifications
CHST15 | |
Polyclonal | |
Unconjugated | |
CHST15 | |
4631426J05Rik; B cell RAG associated protein; B cell RAG associated protein (GALNAC4S-6ST); B-cell RAG-associated gene protein; Brag; carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15; carbohydrate sulfotransferase 15; Chst15; DKFZp781H1369; GalNAc 4-sulfate 6-O-sulfotransferase; GALNAC4S6ST; GALNAC4S-6ST; GalNAcS-6ST; hBRAG; KIAA0598; MAd5; MGC34346; mKIAA0598; N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase; RP11-47G11.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51363 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.94 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q7LFX5 | |
CHST15 | |
Recombinant fragment corresponding to a region within amino acids 203 and 420 of Human CHST15. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction