Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHST2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572125
Description
N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate (PAPS) to position 6 of a nonreducing N-acetylglucosamine (GlcNAc) residue (Uchimura et al., 1998 [PubMed 9722682]).
Specifications
CHST2 | |
Polyclonal | |
Unconjugated | |
CHST2 | |
C6ST; GST-2; GST2; Gn6ST-1; glcNAc6ST-1; N-acetylglucosamine 6-O-sulfotransferase 1; carbohydrate (chondroitin 6/keratan) sulfotransferase 2; carbohydrate sulfotransferase 2; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2 | |
Rabbit | |
Protein A | |
RUO | |
54371, 9435 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.52 mg/mL | |
PBS with 0.09% sodium azide | |
Q80WV3, Q9Y4C5 | |
CHST2 | |
KLH conjugated synthetic peptide between 493-522 amino acids from the C-terminal region of human CHST2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction