Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHTF8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549525

Catalog No. PA549525

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Potential tumor suppressor. Inhibits prostate tumor cell growth, when overexpressed.
Specifications

Specifications

CHTF8
Polyclonal
PBS with 0.09% sodium azide
P0CG12
Chromosome transmission fidelity protein 8 homolog isoform 2, Decreased expression in renal and prostate cancer protein, CHTF8, DERPC
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54921
Western Blot
Unconjugated
CHTF8
Liquid
CHTF8
KLH conjugated synthetic peptide between 2-33 amino acids from human CHTF8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit