Chums™ 1/2 in. 2-ply Cotton Breakaway Lanyards

Comfortable half inch two-ply cotton knit lanyard

Manufacturer: Chums Ltd 11090102

 View more versions of this product

Catalog No. 19-808-7054

  Check Availability
Add to Cart

    Specifications

    Product Type Breakaway Lanyard
    Color Red
    Dimension (Metric) Exterior 24.13cm
    For Use With (Equipment) RHook2 or RHook4
    Material Half inch 2-ply Cotton
    Size (English) 9.5 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit