Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chums™ 13mm Polyester Breakaway Lanyards with Swivel J-Hook

Manufacturer:  Chums™ 11096101

 View more versions of this product

Catalog No. 19-808-7063


Add to cart

Description

Description

  • Reconnecting breakaway pulls apart with six pounds of pressure
  • Swivel J-hook included
  • Available in Black, Royal Blue and Red
  • Logo option available

Specifications

Specifications

Breakaway Lanyard
Safety
1.3cm
Polyester
0.51 in.
Royal
Swivel J-hook
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit