Chums™ 3mm Nylon Rope Breakaway Eyewear Retainers

Field-tested with a reconnecting breakaway for safety

Manufacturer: Chums Ltd 13009101

 View more versions of this product

Catalog No. 19-808-7024

  Check Availability
Add to Cart

    Specifications

    Material Nylon 3mm perlon
    Type Breakaway Retainer
    Color Royal
    For Use With (Equipment) fits a variety of frames
    Includes Single breakaway

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit