Chums™ 3mm Universal Fit Nylon Rope Eyewear Retainers
Durable UV- and water-resistant nylon offers high resistance to solvents, alkalis, acids, abrasion and rot
Manufacturer: Chums Ltd 12103105
 View more versions of this product
Catalog No. 19-808-7021
  Check Availability
Add to Cart
    Specifications
    Material Nylon 3mm perlon
    Type Universal Fit Retainer
    Color Navy
    Includes Adjustable slider bead
  • Description & Specifications
Related Products
Product Content Feedback