Fisher Scientific
Fisher Healthcare
Fisher Science Education
Sign Up for Email
Help & Support Center ›
0
No offer available
Chemical Structure
Certificates
Safety Data Sheets
Order Status
Special Offers & Programs
Order By Catalog Number
Shop Products
Antibodies & Protein Biology
Antibodies & Protein Biology
Antibody Production and Purification
Electrophoresis, Western Blotting and ELISA
Precast Electrophoresis Gels
Primary Antibodies
Secondary Antibodies
View All Antibodies
Biochemicals and Reagents
Biological Buffers
Custom Services and Products
Enzymes and Inhibitors
View All Protein Biology
View All Life Sciences
Balances, Scales & Weighing
Balances, Scales & Weighing
Balance Accessories
Calibration Weights
Laboratory Balances
Microbalances
Scales
Weighing Dishes and Papers
Cell Culture & Analysis
Cell Culture & Analysis
Cell Culture Dishes
Cell Culture Microplates
Cell Culture Flasks
Cell Culture Media
Cryogenic Storage
Fetal Calf and Other Sera
Serological Pipets
View All Cell Culture
Bioprocess Systems and Accessories
Cell Based Assays
Flow Cytometry Products
Microscopes
Cellular Imaging
View All Cell Analysis
View All Life Sciences
Custom DNA Oligos, RNAi & Assays
R&D Systems™ Luminex™ Assays Tool
Centrifuges & Microcentrifuges
Centrifuges & Microcentrifuges
Benchtop Centrifuges
Centrifugal Filter Devices
Centrifuge Accessories
Centrifuge Adapters
Centrifuge Buckets
Floor Model Centrifuges
Microcentrifuges
Rotors
Tubes and Bottles
Chromatography
Chromatography
Autosampler Vial Crimpers and Decappers
Autosampler Vials, Caps, and Closures
Chromatography and Mass Spectrometry Reagents
Chromatography Columns and Cartridges
Chromatography Syringes
Chromatography Water
Gases and Gas Accessories
Paper Chromatography Products
Solid Phase Extraction Products
TLC Products
Chemicals
Chemicals
Acids
Analytical and Chromatography
Bases
Biochemicals and Diagnostics
Heterocyclic Compounds
Chemical Substructure Search
Organic Compounds
Organometallics
Salts and Inorganics
Buffers and Standards
Solvents
Equipment & Instruments
Equipment & Instruments
Balances, Scales and Weighing
Microscopes
pH Meters
Pumps and Tubing
Refractometers
Shakers
Refrigerators and Cold Storage
Rockers and Rotators
Stirrers and Stir Bars
Thermometers
Timers
Forceps, Spatulas & Utensils
Forceps, Spatulas & Utensils
Clamps and Supports
Dissection Equipment
Surgical Tools
Samplers
Scissors and Chisels
Spatulas
Stir Bars and Rods
Medical Syringes
Tongs
Forceps and Tweezers
Furniture, Storage, Carts & Hoods
Furniture, Storage, Carts & Hoods
Boxes and Shipping Supplies
Chairs and Stools
Carts
Desks and Workstations
Laboratory Hoods and Enclosures
Safety Cabinets
Shelving
Storage Cabinets
Wiper Dispensers
Desks and Tables
Glassware & Plasticware
Glassware & Plasticware
Beakers and Lids
Bottles, Jars and Jugs
Carboys
Clamps, Stands and Supports
Coverslips
Dishes
Microplates
Racks for Tubes and Vials
Dispensers
Tubes
View All Lab Consumables
Heating & Cooling
Heating and Cooling
Burners and Lighters
Heaters and Heating Mantles
Hotplates and Hotplate Stirrers
Incubators
Ovens
View All Heating Equipment
Freezers
Liquid Nitrogen Storage Equipment
Refrigerators
Refrigerator and Freezer Accessories
Insulated Containers
View All Cold Storage Products
Influenza, Virus, and COVID-19 Testing
Flu and Virus Testing
COVID Testing Kits
Multiplex COVID Testing Kits
Influenza Testing
Respiratory Viral Testing
PCR & Molecular Biology
PCR & Molecular Biology
Thermal Cyclers
PCR and qPCR Reagents and Kits
PCR Tubes
PCR Plates
Oligonucleotides
RNAi and RNA Reagents
DNA Extraction and Purification
View All PCR
Real-Time PCR Master Mixes
Biochemical Reagents
Life Science Buffers
View All Molecular Biology
View All Life Sciences
Oligos and RNAi Tools
Eurofins Genomics Tool
Invitrogen™ Custom DNA Oligos Tool
Invitrogen™ Custom RNAi Tool
pH & Electrochemistry
pH & Electrochemistry
pH Paper and other Test Strips
Combination Ion Selective Electrodes
Dissolved Oxygen and BOD Probes
Electrochemistry
pH Electrodes and Accessories
Micro pH Electrodes
pH Meters
Conductivity Meters
Dissolved Oxygen Meters
Pipets, Pipettes, Syringes & Needles
Pipets, Pipettes, Syringes & Needles
Serological Pipets
Volumetric Pipets
Pipettes
Pipette Tips and Racks
Dispensers
All Pipets, Pipetters and Tips
Laboratory Syringes
Laboratory Syringe Needles and Accessories
Syringe Filters
Sharps Containers
View All Lab Consumables
Safety, Gloves, Glasses & Cleaning
Safety, Gloves, Glasses & Cleaning
Autoclave Bags
Biohazard Bags
Cleaning Supplies
Disposable Exam Gloves
First Responder Products
Glasses, Goggles, and Face Protection
Hazardous Materials Storage
Lab Coats, Aprons, and Other Safety Apparel
Sharps Disposal Containers and Accessories
Wipes, Cleaners, and Disinfectants
Absorbent Pads and Bench Protectors
Teaching and Education Supplies
Teaching and Education Supplies
Biology Classroom
Biotechnology
Chemistry Classroom
Classroom Furniture
Classroom Laboratory Supplies and Consumables
Classroom Safety
Classroom Science Lab Equipment
Environmental Science
Physics Classroom
STEM and Career Education
Testing & Filtration
Testing and Filtration
Bottle Tops and Filter Units
Extraction Thimbles
Filter Holders and Hardware
Filter Paper
Filtering Funnels
Food and Beverage Testing
Glass Fiber Filters and Prefilters
Membranes for Filtration
Syringe and Syringeless Filters
Water Testing and Environmental Analysis
Tubes & Vials
Tubes and Vials
Centrifuge Tubes
Microcentrifuge Tubes
PCR Tubes
Test Tubes
Tube Racks and Storage
Autosampler Vials, Caps, and Closures
Cryogenic Storage Vials
Sample Vials
Scintillation Vials
Vial Racks and Storage
View All Lab Consumables
RNAi, Oligos, & Assay Tools
RNAi, Oligos, & Assay Tools
Oligos Tools
Eurofins Genomics Tool
Invitrogen™ Custom DNA Oligos Tool
Invitrogen™ Custom RNAi Tool
Assay Tools
Applied Biosystems™ TaqMan® Assay and Arrays Search Tool
Applied Biosystems™ TaqMan® Custom Assay Design Tools
Applied Biosystems™ Custom qPCR Primers and TaqMan® Probes Tool
R&D Systems™ Luminex™ Assays Tool
Chemical Structure Tools
eMolecules Chemical Search Platform
See All Categories
Sign In
My
Account
My Lists
Account Profile
Order By Catalog Number
Register for an Account
Sign In
Please sign in to view account pricing and product availability.
Sign In
Don't have a profile?
Register
0
Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Home
Products
Personal Protective Equipment
Eye Protection and Face Protection
Replacement Lenses
Chums™ Eyeglass Inserts
Print
Chums™ Eyeglass Inserts
Click to view available options
1.50
1.75
Personal prescription use
Description
Documents
Accessories