Learn More
Chums™ Fire Resistant Eyewear Retainer
Will not ignite, smolder, melt or drip when exposed to arc flash, direct flame, sparks or welding spatter
Manufacturer:  Chums™ 12215
 View more versions of this product
Catalog No. 19-808-7040
Add to cart
Description
Description
  • PBI/Kevlar material
  • NFPA 70E Certified
  • Cut and puncture resistant
  • Adjustable non-conductive PVC slider bead
  • Integrity and suppleness retained up to 540C, LOI = 41-38%
  • Fits most safety eyewear
  • Made in the USA
Compliance
NFPA 70E
Specifications
Specifications
Standard end
Fits most safety eyewear
PBI/Dupont™ Kevlar™ aramid fiber
Safety
Adjustable bead