Chums™ Original Cotton Large End Eyewear Retainers

Designed to fit thicker, wider, larger frames

Manufacturer: Chums Ltd 12119108ISF

 View more versions of this product

Catalog No. 19-808-658

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Material Cotton
    Type Large End Retainer
    Color Yellow
    For Use With (Equipment) Suitable for extra-wide and irregular-shaped frames; Compatible with many OTG faceshield
    Includes Adjustable slider bead

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit