Chums™ Original Cotton Single Breakaway Eyewear Retainers

Features a reconnecting single breakaway for situations where choking or snagging hazards exist

Manufacturer: Chums Ltd 13002100

 View more versions of this product

Catalog No. 19-808-7033

  Check Availability
Add to Cart

    Specifications

    Material Cotton
    Type Breakaway Retainer
    Color Black
    For Use With (Equipment) Standard frames
    Includes Reconnecting single breakaway

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit