Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chums™ Original Cotton Standard End Eyewear Retainers
Click to view available options
Color:
Assorted in Pack of 25
Black
Marine Blue-Green
Navy
Orange
Purple
Rainbow Tie-Dye
Red
Royal
Sky Dye-Blue
Description
World's first 100% cotton, fully adjustable eyewear retainer
Tough and durable, yet soft enough for everyday use.
- Fits most standard frames
- Logo option available
- Made in the USA
Order Info
Available in a variety of popular colors.
Specifications
Color | Orange |
Product Type | Eyewear retainer |
For Use With (Equipment) | Fits most standard frames |
Material | Cotton |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction