Chums™ Original Cotton Standard End Eyewear Retainers

World's first 100% cotton, fully adjustable eyewear retainer

Manufacturer: Chums Ltd 12115105ICI

 View more versions of this product

Catalog No. 19-037-626

  Check Availability
Add to Cart

    Specifications

    Material Cotton
    Type Eyewear Retainer
    Color Black with ICI logo
    For Use With (Equipment) Fits most standard frames

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit