Learn More
Abnova™ Chymotrypsin Alpha (Bovine), 1X
Used for Enzyme
Manufacturer:  Abnova™ P5209
Catalog No. 89-939-609
Add to cart
Description
Description
#REF!
Specifications
Specifications
P00766
Lyophilized
Chymotrypsin Alpha (Bovine), 1x
1 g
RUO
Chymotrypsin Assay; Sequence analysis; Peptide synthesis; Peptide mapping; Peptide fingerprinting.
Bovine
Native protein purified from Bovine Pancreas
Enzyme Activity
25.6kDa
Native protein purified from Bovine Pancreas
Store at 4°C.
CHYMOTRYPSIN
≥35 units per mg protein. One Unit hydrolyzes one micromole of benzoyl-L-tyrosine ethyl ester per minute at 25°C, pH 7.8 in the presence of calcium. An activity of 45 units per mg using the above definition, is the equivalent of 10,000 optical density or 1330 N.F. units per mg using ATEE as a substrate.
1 BTEE unit = 29.5 USP/NF units.
None