Learn More
Thermo Scientific Chemicals Chymotrypsinogen A, from bovine pancreas
Description
Chymotrypsinogen A is used as a serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met) on the carboxyl end of the peptide bond, used in the non-invasive determination of solid-state protein conformation using near infrared (NIR) spectroscopy. It has been used to study the partitioning of protein in polymer/polymer aqueous two-phase systems. The enzyme has also been used for self-interaction chromatography applications, to test the rapid measurement of protein osmotic second virial coefficients. In this technique, the protein is immobilized on chromatographic particles and its retention is measured using isocratic elution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Description | White to Off-white |
For Use With (Application) | The zymogen of chymotrypsin |
Molecular Weight (g/mol) | 25.6 kd |
Content And Storage | -30°C to -10°C |
Form | Powder |
Quantity | 5 g |
Product Type | Protease |
Safety and Handling
EINECSNumber | 232-905-3 |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.