Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chymotrypsinogen A, from bovine pancreas

The zymogen of chymotrypsin

$112.00 - $374.00

Specifications

CAS 9035-75-0
Molecular Weight 25.6 kd
Storage Requirements −30° to −10°C
Format Powder
For Use With (Application) The zymogen of chymotrypsin
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6474103 View Documents Alfa Aesar™
J6474103
1g Each for $112.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6474106 View Documents Alfa Aesar™
J6474106
5g Each for $374.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

9035-75-0
−30° to −10°C
The zymogen of chymotrypsin
Protease
25.6 kd
Powder
White to Off-white
Safety and Handling

Safety and Handling

EINECSNumber : 232-905-3

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit