Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CI 976, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
Selective acyl-coenzyme A:cholesterol acyltransferase (ACAT) inhibitor (IC50 = 0.073 μM). Lowers non-high density lipoprotein (HDL)-cholesterol and increases HDL-cholesterol concentrations in rats with pre-established dyslipidemia. Orally active. Also inhibits Golgi-associated LPL acyltransferase (LPAT) activity (IC50 = 15 μM).
Specifications
Quantity | 10 mg |
Formula Weight | 393.57 |
Percent Purity | >98% |
Chemical Name or Material | CI 976 |
Safety and Handling
Recommended Storage | Store at +4°C |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'