Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIAO1 Antibody (6C9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242631AF532
Description
CIAO1 Monoclonal specifically detects CIAO1 in Human samples. It is validated for Western Blot, ELISA.Specifications
CIAO1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
CIAO1 | |
Recombinant human CIAO1 (1-339aa) purified from E. coli | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA | |
6C9 | |
Western Blot, ELISA | |
CIA1, cytosolic iron-sulfur protein assembly 1, cytosolic iron-sulfur protein assembly 1 homolog (S. cerevisiae), probable cytosolic iron-sulfur protein assembly protein CIAO1, WD repeat domain 39, WD repeat-containing protein 39, WD40 protein Ciao1, WDR39cytosolic iron-sulfur protein assembly 1 homolog | |
Mouse | |
0.1 mL | |
9391 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction