Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-1/HIAP-2 Mouse anti-Human, DyLight 680, Clone: 422, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243821FR

Catalog No. NBP243821FR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

cIAP-1/HIAP-2
422
Western Blot
Purified
BIRC2
Recombinant protein encompassing a sequence within the center region of human cIAP1. The exact sequence is proprietary.
Protein G purified
Apoptosis
Primary
329
Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit