Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cIAP-2/HIAP-1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI1A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270184AF700

Catalog No. N270184A700


Add to cart

Description

Description

cIAP-2/HIAP-1 Monoclonal antibody specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

cIAP-2/HIAP-1
OTI1A9
Western Blot, Flow Cytometry
Affinity Purified, Conjugated
BIRC3
Recombinant protein fragment corresponding to amino acids 255-529 of human cIAP-2/HIAP-1 (NP_001156) produced in E.coli.
Immunogen affinity purified
Apoptosis, Cancer, Inhibitors Activators and Regulators, Zinc Finger
Primary
330.0
Flow Cytometry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit