Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIAPIN1 Mouse anti-Human, Clone: OTI2B3, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA808768

Description
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.Specifications
CIAPIN1 | |
Monoclonal | |
Unconjugated | |
CIAPIN1 | |
Anamorsin, DRE2, PRO0915 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57019 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI2B3 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q6FI81 | |
CIAPIN1 | |
Human recombit protein fragment corresponding to amino acids 97-312 of human CIAPIN1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title