Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIB1 Mouse anti-Human, Alexa Fluor 647, Clone: 1D1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104279AF647
Description
CIB1 Monoclonal specifically detects CIB1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CIB1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CIB1 | |
Recombinant human CIB1 (1-191aa) purified from E. coli | |
0.1 mL | |
Signal Transduction | |
10519 | |
Human | |
Purified |
Western Blot, ELISA | |
1D1 | |
Western Blot, ELISA | |
calcium and integrin binding 1 (calmyrin), Calcium- and integrin-binding protein, calmyrin, CIBcalcium and integrin binding protein, CIBP, DNA-dependent protein kinase interacting protein, DNA-PKcs-interacting protein, Kinase-interacting protein, KIP1, KIPcalcium and integrin binding, protein kinase interacting protein, PRKDCIP, SIP2-28calcium and integrin-binding protein 1, Snk interacting protein 2-28, SNK-interacting protein 2-28 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human CIB1 mAb, clone 1D1, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CIB1 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title