Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIB2 Antibody (12), FITC , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305870F
Description
CIB2 Monoclonal antibody specifically detects CIB2 in Human samples. It is validated for ELISASpecifications
CIB2 | |
Monoclonal | |
FITC | |
2810434I23Rik, calcium and integrin binding family member 2, DNA-dependent protein kinase catalytic subunit-interacting protein 2, Kinase-interacting protein 2, KIP 2, KIP2calcium and integrin-binding family member 2 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CIB2 (O75838; Met1-Ile187). | |
0.1 mL | |
Cancer | |
10518 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
12 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction